Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: VASN | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_138440.2 | NP_612449.2 | 2 | 332 | ||
GenBank mRNA | AY166584.1 | - | 1 | 177 | ||
AY358298.1 | - | 2 | 313 | |||
AY358299.1 | - | 2 | 296 | |||
BC068575.1 | - | 2 | 332 | 30349094 |