Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NT5C3B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_052935.4 | NP_443167.4 | 9 | 872 | ||
NR_033464.1 | 9 | 940 | ||||
NR_033465.1 | 8 | 1096 | ||||
XM_006721669.3 | XP_006721732.1 | 10 | 1065 | |||
XM_006721670.3 | XP_006721733.1 | 10 | 956 | |||
XM_011524276.2 | XP_011522578.1 | 8 | 676 | |||
XM_011524277.2 | XP_011522579.1 | 8 | 748 | |||
XM_017024127.1 | XP_016879616.1 | 8 | 714 | |||
GenBank mRNA | BC013742.1 | - | 8 | 917 | 4470687 | |
BC014132.2 | - | 8 | 1021 | 4621938 | ||
BC016971.1 | - | 8 | 1041 | 3885455 | ||
BC067788.1 | - | 8 | 1030 | 5298548 | ||
BF339623.1 | - | 3 | 333 | |||
BX365588.2 | - | 9 | 901 |