Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC26A7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001282356.1 | NP_001269285.1 | 10, 11 | 1428 | ||
NM_001282357.1 | NP_001269286.1 | 9, 10 | 1265 | |||
NM_052832.3 | NP_439897.1 | 9, 10 | 1389 | |||
NM_134266.1 | NP_599028.1 | 9, 10 | 1389 | |||
GenBank mRNA | AF331521.1 | - | 9, 10 | 1359 | ||
AJ413228.1 | - | 9, 10 | 1358 | |||
AJ413229.2 | - | 9, 10 | 1390 | |||
AJ413230.1 | - | 9, 10 | 1358 | |||
AK122933.1 | - | 2, 3 | 280 | |||
BC113866.1 | - | 8, 9 | 1241 | 40034306 | ||
BC114474.1 | - | 8, 9 | 1229 | 40034303 |