Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZNF554 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001102651.1 | NP_001096121.1 | 5 | 1364 | ||
XM_006722635.3 | XP_006722698.1 | 4 | 1051 | |||
XM_011527660.2 | XP_011525962.1 | 5 | 1205 | |||
XM_011527662.2 | XP_011525964.1 | 4 | 1107 | |||
XM_011527664.2 | XP_011525966.1 | 3 | 2336 | |||
XM_017026234.1 | XP_016881723.1 | 5 | 1278 | |||
XM_017026235.1 | XP_016881724.1 | 3 | 2681 | |||
XM_017026236.1 | XP_016881725.1 | 3 | 1049 | |||
GenBank mRNA | AK027860.1 | - | 2 | 2660 | ||
AK092136.1 | - | 6 | 1416 | |||
AK299752.1 | - | 5 | 1270 | |||
BC000113.1 | - | 5 | 1275 | |||
BC047777.1 | - | 5 | 1333 | 6187000 |