Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PCMTD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001286783.1 | NP_001273712.1 | 2 | 277 | ||
NM_052937.3 | NP_443169.2 | 3 | 679 | |||
XM_005251146.2 | XP_005251203.1 | 3 | 679 | |||
XM_011517444.2 | XP_011515746.1 | 2 | 268 | |||
XM_017013017.1 | XP_016868506.1 | 2 | 320 | |||
GenBank mRNA | AK056952.1 | - | 3 | 529 | ||
AK304332.1 | - | 2 | 61 | |||
AL833027.1 | - | 2 | 80 | |||
BC032670.1 | - | 2 | 424 | 5539318 | ||
BP418683.1 | - | 2 | 78 |