Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FCRL3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001320333.1 | NP_001307262.1 | 7 | 1349 | ||
NM_052939.3 | NP_443171.2 | 7 | 1334 | |||
NR_135214.1 | 7 | 1349 | ||||
NR_135215.1 | 7 | 1349 | ||||
NR_135216.1 | 7 | 1254 | ||||
NR_135217.1 | 6 | 1064 | ||||
XM_006711145.1 | XP_006711208.1 | 7 | 1264 | |||
GenBank mRNA | AF416901.1 | - | 7 | 1213 | ||
AF416902.1 | - | 6 | 928 | |||
AF416903.1 | - | 7 | 1213 | |||
AF416904.1 | - | 6 | 1447 | |||
AF416905.1 | - | 7 | 1118 | |||
AF459027.1 | - | 7 | 1334 | |||
AK098122.1 | - | 7 | 1264 | |||
AK301257.1 | - | 6 | 1470 | |||
AL833391.1 | - | 1 | 1884 | |||
AY043466.1 | - | 6 | 1138 | |||
BC028933.1 | - | 7 | 1215 | 4340939 |