Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZNF689 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_138447.2 | NP_612456.1 | 2 | 651 | ||
NR_073480.1 | 2 | 286 | ||||
NR_073481.1 | 2 | 289 | ||||
NR_073482.1 | 2 | 169 | ||||
NR_104166.1 | 2 | 188 | ||||
GenBank mRNA | AB065282.1 | - | 2 | 639 | ||
AK074896.1 | - | 2 | 374 | |||
AK310459.1 | - | 2 | 208 | |||
BC014000.2 | - | 2 | 626 | 3936339 | ||
CN406120.1 | - | 2 | 300 | |||
DA010230.1 | - | 2 | 290 | |||
DA485303.1 | - | 2 | 286 | |||
DB456506.1 | - | 2 | 169 | |||
HY005770.1 | - | 2 | 183 |