Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ARHGEF25 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001111270.2 | NP_001104740.1 | 14, 15 | 1577 | ||
NM_182947.3 | NP_891992.2 | 13, 14 | 1883 | |||
NR_046223.1 | 14, 15 | 1913 | ||||
GenBank mRNA | AB370196.1 | - | 14, 15 | 1577 | ||
AF487514.2 | - | 13, 14 | 1883 | |||
BC012860.1 | - | 14, 15 | 1435 | 4179764 | ||
BC018536.1 | - | 1, 2 | 122 | |||
BC047559.1 | - | 13, 14 | 1883 | 5311038 |