Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC5A11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001258411.2 | NP_001245340.1 | 5 | 601 | ||
NM_001258413.2 | NP_001245342.1 | 4 | 442 | |||
NM_001258414.1 | NP_001245343.1 | 4 | 442 | |||
NM_052944.4 | NP_443176.2 | 4 | 452 | |||
XM_005255080.2 | XP_005255137.1 | 5 | 480 | |||
XM_006721015.2 | XP_006721078.1 | 4 | 565 | |||
XM_011545725.2 | XP_011544027.2 | 5 | 728 | |||
XM_017022900.1 | XP_016878389.1 | 5 | 796 | |||
XM_017022901.1 | XP_016878390.1 | 5 | 1506 | |||
XM_017022902.1 | XP_016878391.1 | 5 | 635 | |||
XM_017022903.1 | XP_016878392.1 | 6 | 645 | |||
XM_017022904.1 | XP_016878393.1 | 6 | 815 | |||
XM_017022905.1 | XP_016878394.1 | 5 | 796 | |||
XM_017022906.1 | XP_016878395.1 | 5 | 656 | |||
XM_017022907.1 | XP_016878396.1 | 5 | 480 | |||
XM_017022908.1 | XP_016878397.1 | 5 | 539 | |||
XM_017022909.1 | XP_016878398.1 | 5 | 480 | |||
GenBank mRNA | AF292385.1 | - | 3 | 220 | ||
AJ305237.1 | - | 3 | 238 | |||
AK125267.1 | - | 6 | 568 | |||
AK298373.1 | - | 3 | 237 | |||
AK303691.1 | - | 5 | 469 | |||
AY044906.1 | - | 4 | 452 | |||
BC049385.1 | - | 4 | 312 | 5755223 | ||
BC057780.1 | - | 4 | 311 | 5287366 |