Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NOSTRIN | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001039724.3 | NP_001034813.2 | 16 | 1491 | ||
NM_001171631.1 | NP_001165102.1 | 21 | 2363 | |||
NM_001171632.1 | NP_001165103.1 | 15 | 1407 | |||
NM_052946.3 | NP_443178.2 | 15 | 1398 | |||
XM_005246270.1 | XP_005246327.1 | 16 | 1550 | |||
XM_011510543.2 | XP_011508845.1 | 10 | 1066 | |||
XM_017003279.1 | XP_016858768.1 | 16 | 1936 | |||
XM_017003280.1 | XP_016858769.1 | 15 | 1413 | |||
GenBank mRNA | AJ532842.1 | - | 16 | 1434 | ||
AK002203.2 | - | 16 | 1463 | |||
AK093444.1 | - | 21 | 2361 | |||
AK123698.1 | - | 5 | 1475 | |||
AK290254.1 | - | 16 | 1457 | |||
BC014189.2 | - | 15 | 1398 | 4650302 | ||
BC093072.1 | - | 16 | 1438 | 30327154 | ||
BF114987.1 | - | 1 | 184 | |||
BQ899074.1 | - | 3 | 180 | |||
DQ402497.1 | - | 15 | 1350 |