Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ALPK2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_052947.3 | NP_443179.3 | 5 | 4037 | ||
XM_011525801.2 | XP_011524103.1 | 5 | 4270 | |||
XM_011525802.2 | XP_011524104.1 | 3 | 4657 | |||
XM_017025535.1 | XP_016881024.1 | 5 | 4271 | |||
XM_017025536.1 | XP_016881025.1 | 5 | 4271 | |||
XM_017025537.1 | XP_016881026.1 | 5 | 4272 | |||
XM_017025538.1 | XP_016881027.1 | 2 | 3155 | |||
XM_017025539.1 | XP_016881028.1 | 2 | 1903 | |||
GenBank mRNA | AK092133.1 | - | 2 | 2155 | ||
AK092194.1 | - | 1 | 756 | |||
AL832018.2 | - | 2 | 2369 | |||
AY044450.1 | - | 2 | 2104 | |||
BX647446.1 | - | 1 | 1515 | |||
BX647639.1 | - | 2 | 2903 | |||
BX647796.1 | - | 5 | 4024 |