Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ARHGAP33 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001172630.1 | NP_001166101.1 | 12 | 1225 | ||
NM_052948.3 | NP_443180.2 | 13 | 1323 | |||
XM_005258488.1 | XP_005258545.1 | 13 | 1319 | |||
XM_006722999.1 | XP_006723062.1 | 13 | 1322 | |||
XM_011526416.1 | XP_011524718.1 | 12 | 1177 | |||
XM_011526417.1 | XP_011524719.1 | 12 | 1180 | |||
XM_011526418.1 | XP_011524720.1 | 12 | 1180 | |||
XM_011526419.1 | XP_011524721.1 | 12 | 1177 | |||
XM_011526420.1 | XP_011524722.1 | 12 | 1228 | |||
XM_011526421.2 | XP_011524723.1 | 12 | 2722 | |||
XM_017026238.1 | XP_016881727.1 | 12 | 2722 | |||
GenBank mRNA | AK096338.1 | - | 13 | 1264 | ||
AK127255.1 | - | 12 | 1225 | |||
BC128586.1 | - | 13 | 1236 | 40129027 |