Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DIS3L | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001143688.2 | NP_001137160.1 | 12 | 1990 | ||
NM_001323936.1 | NP_001310865.1 | 12 | 1909 | |||
NM_001323937.1 | NP_001310866.1 | 11 | 1811 | |||
NM_001323938.1 | NP_001310867.1 | 12 | 1917 | |||
NM_001323939.1 | NP_001310868.1 | 13 | 2114 | |||
NM_001323940.1 | NP_001310869.1 | 11 | 1820 | |||
NM_001323941.1 | NP_001310870.1 | 12 | 2385 | |||
NM_001323943.1 | NP_001310872.1 | 11 | 2014 | |||
NM_001323944.1 | NP_001310873.1 | 12 | 2184 | |||
NM_001323945.1 | NP_001310874.1 | 12 | 2458 | |||
NM_001323946.1 | NP_001310875.1 | 12 | 2111 | |||
NM_001323948.1 | NP_001310877.1 | 11 | 2055 | |||
NM_133375.4 | NP_588616.1 | 12 | 1940 | |||
XM_005254146.3 | XP_005254203.1 | 12 | 2058 | |||
GenBank mRNA | AB075835.1 | - | 11 | 1647 | ||
AK095407.1 | - | 12 | 1969 | |||
AK123202.1 | - | 2 | 2456 | |||
BC014124.2 | - | 4 | 738 | |||
BC022089.2 | - | 12 | 1875 | 3029342 |