Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DIS3L | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001143688.2 | NP_001137160.1 | 4 | 536 | ||
NM_001323936.1 | NP_001310865.1 | 4 | 455 | |||
NM_001323937.1 | NP_001310866.1 | 3 | 357 | |||
NM_001323938.1 | NP_001310867.1 | 4 | 463 | |||
NM_001323939.1 | NP_001310868.1 | 5 | 660 | |||
NM_001323940.1 | NP_001310869.1 | 4 | 536 | |||
NM_001323941.1 | NP_001310870.1 | 4 | 931 | |||
NM_001323943.1 | NP_001310872.1 | 4 | 730 | |||
NM_001323944.1 | NP_001310873.1 | 4 | 730 | |||
NM_001323945.1 | NP_001310874.1 | 4 | 1004 | |||
NM_001323946.1 | NP_001310875.1 | 4 | 657 | |||
NM_001323948.1 | NP_001310877.1 | 3 | 601 | |||
NM_133375.4 | NP_588616.1 | 4 | 486 | |||
XM_005254146.3 | XP_005254203.1 | 4 | 604 | |||
GenBank mRNA | AB075835.1 | - | 3 | 193 | ||
AK095407.1 | - | 4 | 515 | |||
AK295392.1 | - | 4 | 363 | |||
BC022089.2 | - | 4 | 421 | 3029342 | ||
BG722405.1 | - | 4 | 466 | |||
BP219623.1 | - | 4 | 532 |