Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RAB3C | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001317915.1 | NP_001304844.1 | 2 | 370 | ||
NM_138453.3 | NP_612462.1 | 2 | 332 | |||
GenBank mRNA | AY026936.1 | - | 2 | 290 | ||
BC013033.2 | - | 2 | 264 | 3534915 | ||
BX394661.2 | - | 2 | 342 | |||
DB485923.1 | - | 2 | 336 |