Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZNF653 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_138783.3 | NP_620138.2 | 7 | 1610 | ||
GenBank mRNA | AK314986.1 | - | 6 | 1641 | ||
AY072704.1 | - | 7 | 1479 | |||
BC014187.2 | - | 7 | 1506 | 4634585 | ||
BC016816.1 | - | 7 | 1530 | 4094670 |