Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC22A12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001276326.1 | NP_001263255.1 | 9 | 2226 | ||
NM_001276327.1 | NP_001263256.1 | 7 | 2004 | |||
NM_144585.3 | NP_653186.2 | 9 | 2328 | |||
NM_153378.2 | NP_700357.1 | 9 | 2062 | |||
XM_006718430.3 | XP_006718493.1 | 8 | 2009 | |||
XM_006718431.3 | XP_006718494.1 | 9 | 1895 | |||
GenBank mRNA | AB050269.1 | - | 9 | 2325 | ||
AB071863.1 | - | 9 | 1728 | |||
AK309588.1 | - | 7 | 939 | |||
AK315061.1 | - | 9 | 1774 | |||
AY358183.1 | - | 7 | 1537 | |||
BC053348.1 | - | 9 | 1871 | 5183650 |