Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KLHDC3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_057161.3 | NP_476502.1 | 5 | 800 | ||
NR_040101.1 | 4 | 684 | ||||
GenBank mRNA | AB055925.1 | - | 5 | 664 | ||
AK097719.1 | - | 3 | 526 | |||
AK290384.1 | - | 5 | 656 | |||
AK302336.1 | - | 4 | 439 | |||
AK308917.1 | - | 5 | 686 | |||
BC000295.2 | - | 5 | 620 | 2819570 | ||
BC001789.1 | - | 5 | 635 | 3355470 | ||
BC001793.2 | - | 5 | 598 | 3352397 | ||
BC007296.1 | - | 5 | 650 | 3349731 | ||
BC009460.1 | - | 5 | 648 | 3537223 | ||
BC021546.1 | - | 5 | 637 | 3351635 | ||
BC041793.1 | - | 4 | 538 | 5288540 | ||
BC045612.1 | - | 4 | 538 | 5313255 |