Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CYYR1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001320768.1 | NP_001307697.1 | 4 | 916 | ||
NM_052954.3 | NP_443186.2 | 4 | 913 | |||
NR_135472.1 | 5 | 1083 | ||||
GenBank mRNA | AF401639.1 | - | 4 | 518 | ||
AK054581.1 | - | 4 | 793 | |||
AK223576.1 | - | 4 | 633 | |||
AK304124.1 | - | 5 | 963 | |||
AK313231.1 | - | 4 | 699 | |||
AL833200.1 | - | 4 | 724 | |||
AY061853.1 | - | 4 | 458 | |||
BC036761.1 | - | 4 | 701 | 5588436 | ||
KC843341.1 | - | 3 | 353 | |||
KF154022.1 | - | 5 | 685 |