Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ACSM1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001318890.1 | NP_001305819.1 | 3 | 519 | ||
NM_052956.2 | NP_443188.2 | 2 | 383 | |||
NR_134918.1 | 2 | 383 | ||||
XM_006721016.2 | XP_006721079.1 | 2 | 362 | |||
XM_006721017.2 | XP_006721080.1 | 2 | 362 | |||
XM_011545729.2 | XP_011544031.1 | 2 | 362 | |||
XM_011545730.2 | XP_011544032.1 | 2 | 362 | |||
XM_017022914.1 | XP_016878403.1 | 2 | 362 | |||
GenBank mRNA | AB059429.1 | - | 2 | 383 | ||
AK314282.1 | - | 3 | 475 | |||
BC125177.1 | - | 2 | 383 | |||
BC125178.1 | - | 2 | 383 | 40121956 | ||
CN360958.1 | - | 4 | 554 |