Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PANX3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_052959.2 | NP_443191.1 | 4 | 675 | ||
GenBank mRNA | AF406650.1 | - | 4 | 675 |