Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TM4SF18 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001184723.1 | NP_001171652.1 | 4 | 514 | ||
NM_138786.3 | NP_620141.1 | 5 | 690 | |||
GenBank mRNA | AK313400.1 | - | 5 | 464 | ||
AL833134.1 | - | 5 | 612 | |||
BC014339.1 | - | 5 | 566 | 3828466 | ||
BF213446.1 | - | 523 | ||||
CA454331.1 | - | 4 | 476 |