Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TOP1MT | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001258446.1 | NP_001245375.1 | 12 | 1472 | ||
NM_001258447.1 | NP_001245376.1 | 11 | 1331 | |||
NM_052963.2 | NP_443195.1 | 11 | 1380 | |||
XM_005250781.3 | XP_005250838.1 | 11 | 1368 | |||
GenBank mRNA | AF349017.1 | - | 11 | 1369 | ||
AK302372.1 | - | 11 | 1331 | |||
AK303237.1 | - | 10 | 1112 | |||
AK308475.1 | - | 11 | 1353 | |||
AK309005.1 | - | 11 | 1545 | |||
AK311647.1 | - | 8 | 846 | |||
AK315913.1 | - | 11 | 1342 | |||
AK316380.1 | - | 12 | 1419 | |||
BC044646.1 | - | 11 | 1355 | |||
BC052285.1 | - | 11 | 1355 | 5443410 | ||
BC071914.1 | - | 11 | 1353 | 6372930 |