Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TOP1MT | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001258446.1 | NP_001245375.1 | 6 | 642 | ||
NM_001258447.1 | NP_001245376.1 | 5 | 501 | |||
NM_052963.2 | NP_443195.1 | 5 | 550 | |||
XM_005250781.3 | XP_005250838.1 | 5 | 538 | |||
GenBank mRNA | AF349017.1 | - | 5 | 539 | ||
AK302372.1 | - | 5 | 501 | |||
AK309005.1 | - | 5 | 659 | |||
AK315913.1 | - | 5 | 512 | |||
AK316380.1 | - | 6 | 589 | |||
BC044646.1 | - | 5 | 525 | |||
BC052285.1 | - | 5 | 525 | 5443410 | ||
BC071914.1 | - | 5 | 523 | 6372930 | ||
DC375680.1 | - | 5 | 550 | |||
DC399840.1 | - | 5 | 501 |