Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TSEN15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001127394.3 | NP_001120866.1 | 2 | 285 | ||
NM_001300764.1 | NP_001287693.1 | 2 | 285 | |||
NM_001300766.1 | NP_001287695.1 | 2 | 285 | |||
NM_052965.3 | NP_443197.1 | 2 | 285 | |||
NR_023349.2 | 2 | 285 | ||||
NR_125335.1 | 2 | 285 | ||||
XM_006711148.1 | XP_006711211.1 | 2 | 234 | |||
XM_011509139.2 | XP_011507441.1 | 2 | 1729 | |||
XM_017000228.1 | XP_016855717.1 | 2 | 1729 | |||
XM_017000229.1 | XP_016855718.1 | 2 | 1729 | |||
XM_017000230.1 | XP_016855719.1 | 2 | 1729 | |||
GenBank mRNA | AF288394.1 | - | 2 | 260 | ||
AK098196.1 | - | 2 | 183 | |||
AK296655.1 | - | 2 | 180 | |||
BC022030.1 | - | 2 | 230 | 4824025 | ||
BI821111.1 | - | 2 | 210 | |||
BM146150.1 | - | 2 | 287 | |||
BM887891.1 | - | 2 | 117 | |||
BU195337.1 | - | 2 | 257 |