Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HSPA12B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001197327.1 | NP_001184256.1 | 13 | 2182 | ||
NM_001318322.1 | NP_001305251.1 | 12 | 2087 | |||
NM_052970.4 | NP_443202.3 | 13 | 2185 | |||
XM_011529151.2 | XP_011527453.1 | 10 | 1883 | |||
XM_017027632.1 | XP_016883121.1 | 13 | 2100 | |||
GenBank mRNA | AK056712.1 | - | 13 | 2122 | ||
AK297120.1 | - | 11 | 1978 | |||
AK301294.1 | - | 12 | 2053 | |||
BC090857.1 | - | 12 | 2046 | |||
BC110881.1 | - | 13 | 2134 | 5725594 | ||
BC117284.1 | - | 13 | 2083 | 40125835 | ||
BC126263.1 | - | 13 | 2083 | 8991979 | ||
BC143932.1 | - | 13 | 2080 | 9052450 |