Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LRG1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_052972.2 | NP_443204.1 | 2 | 444 | ||
GenBank mRNA | AB073607.1 | - | 2 | 456 | ||
AF403428.1 | - | 2 | 444 | |||
AK027382.1 | - | 2 | 456 | |||
AK314506.1 | - | 2 | 456 | |||
BC034389.1 | - | 2 | 445 | 5187278 | ||
BC070198.1 | - | 2 | 501 | 4692618 |