Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: WDR17 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_170710.4 | NP_733828.2 | 28 | 3844 | ||
NM_181265.3 | NP_851782.3 | 26 | 3676 | |||
XM_005262725.3 | XP_005262782.1 | 26 | 3343 | |||
XM_011531582.2 | XP_011529884.1 | 27 | 3685 | |||
XM_017007686.1 | XP_016863175.1 | 27 | 3673 | |||
XM_017007687.1 | XP_016863176.1 | 28 | 3758 | |||
XM_017007688.1 | XP_016863177.1 | 28 | 3675 | |||
XM_017007689.1 | XP_016863178.1 | 26 | 3651 | |||
XM_017007690.1 | XP_016863179.1 | 27 | 3737 | |||
XM_017007691.1 | XP_016863180.1 | 26 | 3507 | |||
XM_017007692.1 | XP_016863181.1 | 27 | 3593 | |||
XM_017007693.1 | XP_016863182.1 | 25 | 3486 | |||
XM_017007694.1 | XP_016863183.1 | 26 | 3470 | |||
XM_017007695.1 | XP_016863184.1 | 26 | 3521 | |||
XM_017007696.1 | XP_016863185.1 | 25 | 3500 | |||
GenBank mRNA | AF492460.1 | - | 28 | 3697 | ||
AK130128.1 | - | 12 | 1306 | |||
BX648508.1 | - | 12 | 1244 |