Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ACAP3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_030649.2 | NP_085152.2 | 11 | 890 | ||
XM_005244715.2 | XP_005244772.1 | 11 | 926 | |||
XM_011540606.2 | XP_011538908.1 | 11 | 939 | |||
XM_011540607.1 | XP_011538909.1 | 11 | 3363 | |||
XM_011540608.2 | XP_011538910.1 | 10 | 844 | |||
XM_011540609.2 | XP_011538911.1 | 11 | 940 | |||
XM_017000233.1 | XP_016855722.1 | 11 | 1079 | |||
GenBank mRNA | AB051503.1 | - | 10 | 2555 | ||
AF411981.1 | - | 9 | 690 | |||
BC047001.1 | - | 1 | 342 | |||
BC051194.1 | - | 11 | 890 | 4819254 |