Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ARAP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015230.3 | NP_056045.2 | 26 | 4468 | ||
XM_017007698.1 | XP_016863187.1 | 26 | 4898 | |||
XM_017007699.1 | XP_016863188.1 | 26 | 4353 | |||
XM_017007700.1 | XP_016863189.1 | 25 | 4829 | |||
XM_017007701.1 | XP_016863190.1 | 26 | 4898 | |||
XM_017007702.1 | XP_016863191.1 | 25 | 4829 | |||
GenBank mRNA | AB011152.2 | - | 25 | 3911 | ||
AF411982.1 | - | 15 | 1907 | |||
AF439781.1 | - | 25 | 4290 | |||
AY049733.1 | - | 26 | 4383 | |||
BC105089.1 | - | 13 | 1682 | |||
BC105091.1 | - | 13 | 1682 | |||
BC150258.1 | - | 25 | 3922 |