Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ARAP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015230.3 | NP_056045.2 | 26, 27 | 4504 | ||
XM_017007698.1 | XP_016863187.1 | 26, 27 | 4934 | |||
XM_017007699.1 | XP_016863188.1 | 26, 27 | 4389 | |||
XM_017007700.1 | XP_016863189.1 | 25, 26 | 4865 | |||
XM_017007701.1 | XP_016863190.1 | 26, 27 | 4934 | |||
XM_017007702.1 | XP_016863191.1 | 25, 26 | 4865 | |||
GenBank mRNA | AB011152.2 | - | 25, 26 | 3947 | ||
AF411982.1 | - | 15, 16 | 1943 | |||
AF439781.1 | - | 25, 26 | 4326 | |||
AY049733.1 | - | 26, 27 | 4419 | |||
BC105089.1 | - | 13, 14 | 1718 | |||
BC105091.1 | - | 13, 14 | 1718 | |||
BC150258.1 | - | 25, 26 | 3958 |