Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TADA1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053053.3 | NP_444281.1 | 5 | 614 | ||
GenBank mRNA | AK290964.1 | - | 5 | 431 | ||
AK291922.1 | - | 5 | 554 | |||
AK310391.1 | - | 1 | 287 | |||
BC015401.2 | - | 5 | 439 | 4182025 |