Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SCGB3A2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_054023.4 | NP_473364.1 | 2 | 220 | ||
GenBank mRNA | AA988559.1 | - | 2 | 297 | ||
AF313455.1 | - | 2 | 141 | |||
AF439545.1 | - | 2 | 209 | |||
AY358979.1 | - | 2 | 209 | |||
BC024232.1 | - | 2 | 224 | 4694514 |