Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FTSJ3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017647.3 | NP_060117.3 | 8 | 1325 | ||
GenBank mRNA | AF327355.1 | - | 7 | 800 | ||
AK095861.1 | - | 8 | 1325 | |||
AK296715.1 | - | 7 | 531 | |||
AK307398.1 | - | 7 | 814 | |||
AK315010.1 | - | 8 | 750 | |||
AL834482.1 | - | 2 | 213 | |||
BC000131.1 | - | 3 | 340 | |||
BC036710.2 | - | 7 | 709 | 4641078 |