Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TMC2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_080751.2 | NP_542789.2 | 9 | 1065 | ||
XM_005260660.3 | XP_005260717.1 | 7 | 1259 | |||
GenBank mRNA | AF417580.2 | - | 9 | 1065 | ||
AK294013.1 | - | 7 | 811 | |||
AK316261.1 | - | 7 | 781 |