Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TMC2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_080751.2 | NP_542789.2 | 11 | 1293 | ||
XM_005260660.3 | XP_005260717.1 | 9 | 1487 | |||
GenBank mRNA | AF417580.2 | - | 11 | 1293 | ||
AK294013.1 | - | 9 | 1039 | |||
AK316261.1 | - | 9 | 1009 |