Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: UBE2J2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_058167.2 | NP_477515.2 | 4 | 438 | ||
NM_194315.1 | NP_919296.1 | 5 | 486 | |||
NM_194457.1 | NP_919439.1 | 3 | 307 | |||
NM_194458.1 | NP_919440.1 | 4 | 567 | |||
XM_005244718.3 | XP_005244775.1 | 4 | 716 | |||
XM_005244719.4 | XP_005244776.1 | 4 | 604 | |||
XM_006710333.3 | XP_006710396.1 | 4 | 883 | |||
XM_011540613.2 | XP_011538915.1 | 3 | 311 | |||
XM_011540614.2 | XP_011538916.1 | 4 | 669 | |||
XM_011540616.2 | XP_011538918.1 | 5 | 678 | |||
XM_011540617.2 | XP_011538919.1 | 4 | 445 | |||
XM_017000238.1 | XP_016855727.1 | 5 | 549 | |||
XM_017000239.1 | XP_016855728.1 | 3 | 767 | |||
GenBank mRNA | AF296658.1 | - | 4 | 391 | ||
AK056065.1 | - | 4 | 348 | |||
AK075017.1 | - | 4 | 417 | |||
AK291935.1 | - | 4 | 417 | |||
AL555293.3 | - | 5 | 562 | |||
BC094777.1 | - | 4 | 437 | 30340049 | ||
BC104890.1 | - | 3 | 262 | 8143893 | ||
BC113430.1 | - | 3 | 262 | 8322482 | ||
BC143657.1 | - | 3 | 262 | 9052171 | ||
BE382927.1 | - | 4 | 381 | |||
BG112933.1 | - | 562 | ||||
BG717336.1 | - | 5 | 664 | |||
BM544827.1 | - | 5 | 489 | |||
BM765278.1 | - | 3 | 222 | |||
BM926567.1 | - | 4 | 564 | |||
BQ881730.1 | - | 2 | 127 |