Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: UBE2J2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_058167.2 | NP_477515.2 | 6, 7 | 679 | ||
NM_194315.1 | NP_919296.1 | 7, 8 | 727 | |||
NM_194457.1 | NP_919439.1 | 5, 6 | 548 | |||
NM_194458.1 | NP_919440.1 | 6, 7 | 808 | |||
XM_005244718.3 | XP_005244775.1 | 6, 7 | 957 | |||
XM_005244719.4 | XP_005244776.1 | 6, 7 | 845 | |||
XM_006710333.3 | XP_006710396.1 | 6, 7 | 1124 | |||
XM_011540613.2 | XP_011538915.1 | 5, 6 | 552 | |||
XM_011540614.2 | XP_011538916.1 | 6, 7 | 910 | |||
XM_011540616.2 | XP_011538918.1 | 7, 8 | 919 | |||
XM_011540617.2 | XP_011538919.1 | 6, 7 | 686 | |||
XM_017000238.1 | XP_016855727.1 | 7, 8 | 790 | |||
XM_017000239.1 | XP_016855728.1 | 5, 6 | 1008 | |||
GenBank mRNA | AF296658.1 | - | 6, 7 | 632 | ||
AK056065.1 | - | 6, 7 | 589 | |||
AK075017.1 | - | 6, 7 | 658 | |||
AK098803.1 | - | 1, 2 | 45 | |||
AK291935.1 | - | 6, 7 | 658 | |||
AL555293.3 | - | 7, 8 | 803 | |||
AW411508.1 | - | 2, 3 | 207 | |||
BC094777.1 | - | 6, 7 | 678 | 30340049 | ||
BC104890.1 | - | 5, 6 | 503 | 8143893 | ||
BC113430.1 | - | 5, 6 | 503 | 8322482 | ||
BC143657.1 | - | 5, 6 | 503 | 9052171 | ||
BM544827.1 | - | 7, 8 | 730 | |||
BM765278.1 | - | 5, 6 | 463 | |||
BM794599.1 | - | 4, 5 | 418 | |||
BQ881730.1 | - | 4, 5 | 368 |