Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: OLFM3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001288823.1 | NP_001275752.1 | 1 | 461 | ||
NM_058170.3 | NP_477518.2 | 1 | 461 | |||
NR_110210.1 | 1 | 461 | ||||
NR_110211.1 | 1 | 461 | ||||
GenBank mRNA | AF397394.1 | - | 1 | 132 | ||
AF397395.1 | - | 1 | 132 | |||
AF397396.1 | - | 1 | 132 | |||
AF397397.1 | - | 1 | 132 | |||
AK095724.1 | - | 1 | 257 | |||
BC022531.1 | - | 1 | 219 | 4812035 | ||
DA406512.1 | - | 1 | 456 |