Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: OLFM3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001288821.1 | NP_001275750.1 | 4 | 613 | ||
NM_001288823.1 | NP_001275752.1 | 5 | 925 | |||
NM_058170.3 | NP_477518.2 | 4 | 864 | |||
NR_110210.1 | 5 | 925 | ||||
NR_110211.1 | 4 | 864 | ||||
NR_110212.1 | 4 | 613 | ||||
XM_017000240.1 | XP_016855729.1 | 5 | 768 | |||
GenBank mRNA | AF397392.2 | - | 4 | 506 | ||
AF397393.2 | - | 4 | 506 | |||
AF397394.1 | - | 4 | 535 | |||
AF397395.1 | - | 4 | 535 | |||
AF397396.1 | - | 5 | 596 | |||
AF397397.1 | - | 5 | 596 | |||
AK095551.1 | - | 2 | 90 | |||
AK095724.1 | - | 4 | 660 | |||
AK300870.1 | - | 4 | 613 | |||
AY358722.1 | - | 4 | 577 | |||
BC022531.1 | - | 4 | 622 | 4812035 |