Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MSS51 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001024593.1 | NP_001019764.1 | 6 | 1188 | ||
GenBank mRNA | AK096884.1 | - | 6 | 758 | ||
AK309870.1 | - | 7 | 1244 | |||
BC094693.1 | - | 6 | 1167 | 30529259 | ||
BC109255.1 | - | 5 | 1101 | |||
BC109256.1 | - | 5 | 1101 | |||
BX537678.1 | - | 5 | 1988 | |||
CR936824.1 | - | 6 | 1170 |