Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CFAP70 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145170.3 | NP_660153.3 | 26 | 3306 | ||
XM_005269489.2 | XP_005269546.1 | 25 | 3176 | |||
XM_006717604.2 | XP_006717667.1 | 27 | 4867 | |||
XM_006717605.2 | XP_006717668.1 | 26 | 3269 | |||
XM_006717609.2 | XP_006717672.1 | 24 | 3083 | |||
XM_006717610.2 | XP_006717673.1 | 25 | 4555 | |||
XM_006717611.3 | XP_006717674.1 | 25 | 3207 | |||
XM_011539212.2 | XP_011537514.1 | 22 | 2838 | |||
XM_017015620.1 | XP_016871109.1 | 27 | 4713 | |||
XM_017015621.1 | XP_016871110.1 | 27 | 4710 | |||
XM_017015622.1 | XP_016871111.1 | 27 | 4710 | |||
XM_017015623.1 | XP_016871112.1 | 27 | 4635 | |||
XM_017015624.1 | XP_016871113.1 | 26 | 4632 | |||
XM_017015625.1 | XP_016871114.1 | 26 | 4623 | |||
XM_017015627.1 | XP_016871116.1 | 26 | 4503 | |||
XM_017015628.1 | XP_016871117.1 | 25 | 3085 | |||
XM_017015629.1 | XP_016871118.1 | 25 | 4308 | |||
XM_017015630.1 | XP_016871119.1 | 26 | 4787 | |||
XM_017015631.1 | XP_016871120.1 | 22 | 2644 | |||
XM_017015632.1 | XP_016871121.1 | 25 | 4682 | |||
GenBank mRNA | AF435958.1 | - | 7 | 830 | ||
AF435959.1 | - | 8 | 920 | |||
AK098631.1 | - | 10 | 1316 | |||
BC032856.2 | - | 26 | 3301 | 5267050 |