Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: C10orf90 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001004298.2 | NP_001004298.2 | 5 | 1672 | ||
XM_005269491.1 | XP_005269548.1 | 5 | 1742 | |||
XM_005269492.2 | XP_005269549.1 | 3 | 1035 | |||
XM_011539216.1 | XP_011537518.1 | 6 | 2400 | |||
XM_011539217.2 | XP_011537519.1 | 6 | 2095 | |||
XM_011539220.2 | XP_011537522.1 | 6 | 1640 | |||
XM_011539222.2 | XP_011537524.1 | 6 | 1762 | |||
XM_011539223.2 | XP_011537525.1 | 5 | 2450 | |||
XM_011539224.2 | XP_011537526.1 | 5 | 1596 | |||
XM_011539226.1 | XP_011537528.1 | 5 | 1595 | |||
XM_011539227.2 | XP_011537529.1 | 5 | 1928 | |||
XM_011539230.2 | XP_011537532.1 | 4 | 1717 | |||
XM_011539231.1 | XP_011537533.1 | 5 | 3727 | |||
XM_011539232.1 | XP_011537534.1 | 5 | 1394 | |||
XM_011539233.1 | XP_011537535.1 | 4 | 1157 | |||
XM_017015634.1 | XP_016871123.1 | 5 | 2109 | |||
GenBank mRNA | AF435960.1 | - | 2 | 179 | ||
AK057500.1 | - | 5 | 1672 | |||
AK297577.1 | - | 6 | 1872 | |||
BC021140.1 | - | 2 | 186 | |||
BC140898.1 | - | 5 | 1659 | 9021766 | ||
BC146869.1 | - | 5 | 1659 | 9056797 |