Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: C10orf90 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001004298.2 | NP_001004298.2 | 3 | 406 | ||
XM_005269491.1 | XP_005269548.1 | 3 | 476 | |||
XM_011539216.1 | XP_011537518.1 | 4 | 1134 | |||
XM_011539217.2 | XP_011537519.1 | 4 | 829 | |||
XM_011539220.2 | XP_011537522.1 | 4 | 374 | |||
XM_011539222.2 | XP_011537524.1 | 4 | 496 | |||
XM_011539223.2 | XP_011537525.1 | 3 | 1184 | |||
XM_011539224.2 | XP_011537526.1 | 3 | 330 | |||
XM_011539226.1 | XP_011537528.1 | 3 | 329 | |||
XM_011539227.2 | XP_011537529.1 | 3 | 662 | |||
XM_011539230.2 | XP_011537532.1 | 1 | 69 | |||
XM_017015634.1 | XP_016871123.1 | 4 | 1134 | |||
XM_017015635.1 | XP_016871124.1 | 4 | 1134 | |||
GenBank mRNA | AK057500.1 | - | 3 | 406 | ||
AK093093.1 | - | 5 | 902 | |||
AK297577.1 | - | 4 | 606 | |||
AK308357.1 | - | 4 | 493 | |||
AK309217.1 | - | 4 | 517 | |||
AL705062.1 | - | 4 | 525 | |||
BC140898.1 | - | 3 | 393 | 9021766 | ||
BC146869.1 | - | 3 | 393 | 9056797 |