Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PDZD8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_173791.4 | NP_776152.1 | 3, 4 | 1332 | ||
XM_005269518.3 | XP_005269575.1 | 2, 3 | 1182 | |||
GenBank mRNA | AK091746.1 | - | 3, 4 | 1305 | ||
AY211929.1 | - | 3, 4 | 401 | |||
BC028375.1 | - | 3, 4 | 1294 | 4837939 |