Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GSTO2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001191014.1 | NP_001177943.1 | 3 | 466 | ||
NM_183239.1 | NP_899062.1 | 5 | 1031 | |||
XM_006717624.3 | XP_006717687.1 | 5 | 720 | |||
XM_011539270.2 | XP_011537572.1 | 5 | 2094 | |||
XM_011539271.1 | XP_011537573.1 | 5 | 720 | |||
XM_011539272.2 | XP_011537574.1 | 5 | 720 | |||
GenBank mRNA | AK291886.1 | - | 5 | 719 | ||
AK300508.1 | - | 4 | 428 | |||
AY191318.1 | - | 4 | 399 | |||
AY209189.1 | - | 5 | 723 | |||
AY350731.1 | - | 5 | 723 | |||
BC023583.2 | - | 3 | 432 | |||
BC056918.1 | - | 5 | 1031 | 5165084 |