Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ANAPC16 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001242546.1 | NP_001229475.1 | 4 | 647 | ||
NM_001242547.1 | NP_001229476.1 | 3 | 255 | |||
NM_173473.3 | NP_775744.1 | 3 | 321 | |||
NR_038391.1 | 2 | 86 | ||||
NR_038392.1 | 2 | 152 | ||||
GenBank mRNA | BC009530.1 | - | 3 | 273 | 3890955 | |
BM768948.1 | - | 2 | 124 | |||
CD511334.1 | - | 3 | 248 | |||
DA310509.1 | - | 2 | 86 | |||
DA739927.1 | - | 3 | 321 | |||
EU552924.1 | - | 3 | 294 |