Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PIH1D2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001082619.1 | NP_001076088.1 | 4 | 735 | ||
NM_138789.3 | NP_620144.1 | 4 | 735 | |||
XM_011542593.2 | XP_011540895.2 | 4 | 641 | |||
XM_017017201.1 | XP_016872690.1 | 4 | 852 | |||
XM_017017202.1 | XP_016872691.1 | 4 | 852 | |||
XM_017017203.1 | XP_016872692.1 | 4 | 852 | |||
XM_017017204.1 | XP_016872693.1 | 4 | 852 | |||
XM_017017205.1 | XP_016872694.1 | 4 | 852 | |||
GenBank mRNA | AK300496.1 | - | 4 | 598 | ||
BC019238.2 | - | 3 | 544 | 3935322 | ||
CA312666.1 | - | 3 | 525 |