Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DEPDC4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001319310.1 | NP_001306239.1 | 4 | 757 | ||
NM_001319311.1 | NP_001306240.1 | 5 | 796 | |||
NM_152317.3 | NP_689530.1 | 4 | 757 | |||
NR_135073.1 | 4 | 757 | ||||
XM_011537876.2 | XP_011536178.1 | 3 | 360 | |||
XM_017018779.1 | XP_016874268.1 | 5 | 796 | |||
XM_017018780.1 | XP_016874269.1 | 4 | 757 | |||
XM_017018781.1 | XP_016874270.1 | 4 | 757 | |||
XM_017018782.1 | XP_016874271.1 | 5 | 689 | |||
XM_017018783.1 | XP_016874272.1 | 5 | 924 | |||
XM_017018784.1 | XP_016874273.1 | 2 | 308 | |||
XM_017018785.1 | XP_016874274.1 | 5 | 796 | |||
GenBank mRNA | AK090824.1 | - | 4 | 757 | ||
BC015117.1 | - | 4 | 657 | |||
BC100927.2 | - | 3 | 360 | |||
BC100928.2 | - | 5 | 595 | |||
BC100929.1 | - | 3 | 360 | |||
BC100930.1 | - | 4 | 757 |