Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LRIG3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001136051.2 | NP_001129523.1 | 10 | 1073 | ||
NM_153377.4 | NP_700356.2 | 10 | 1492 | |||
XM_011537893.2 | XP_011536195.1 | 10 | 1164 | |||
XM_017018790.1 | XP_016874279.1 | 10 | 1494 | |||
GenBank mRNA | AK292857.1 | - | 10 | 1473 | ||
AY358288.1 | - | 10 | 1435 | |||
AY358295.1 | - | 10 | 1073 | |||
AY505340.1 | - | 10 | 1243 | |||
BC126169.1 | - | 10 | 1388 | 8991885 | ||
BC126171.1 | - | 10 | 1388 | 8991887 | ||
BC143960.1 | - | 10 | 1388 |